Learn More
Beckman Coulter Flow-Count Fluorospheres, 200 Tests, CE, IVD
Supplier:  Beckman Coulter 7547053
Encompass_Preferred
Flow-Count Fluorospheres are a suspension of fluorescent microbeads used to determine absolute counts on the flow cytometer. Each fluorosphere contains a dye which has a fluorescent emission range of 525 nm to 700 nm when excited at 488 nm. They have uniform size and fluorescence intensity, and an assayed concentration allowing a direct determination of absolute counts.
Catalog No. NC1908933