Learn More
Medchemexpress LLC Coumachlor | 81-82-3 | 342.77 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW013200500MG
Encompass_Preferred
Coumachlor is an orally active anticoagulant agent and a vitamin K antagonist, intended for research use only.
  • Orally active anticoagulant agent
  • Vitamin K antagonist
  • For research use only
Catalog No. 50-004-54152