Learn More
Medchemexpress LLC Coumalic acid | 500-05-0 | 97.65% | 140.09 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY32004500MG
Encompass_Preferred
Coumalic acid is an inhibitor of human carbonic anhydrase (hCA) with a KI of 0.073 μM for hCA IX and 0.083 μM for hCA XII. It is a valuable platform compound that can be prepared from malic acid. It finds applications in the flavorings, fragrances, and cosmetics industries, as well as in polymer components and pharmaceutical scaffolds. Additionally, it exhibits antibronchial and antimalarial activities.
  • Inhibitor of human carbonic anhydrase (hCA)
  • Valuable platform compound derivable from malic acid
  • Suitable for flavorings, fragrances, and cosmetics industries
  • Used in polymer components
  • Used in pharmaceutical scaffolds
  • Exhibits antibronchial activity
  • Exhibits antimalarial activity
Catalog No. 50-004-01379