Learn More
Medchemexpress LLC Coumalic acid | 500-05-0 | 97.7% | 140.09 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY3200410MM/1ML
Encompass_Preferred
Coumalic acid is an inhibitor of human carbonic anhydrase (hCA) (KI = 0.073 μM for hCA IX; KI = 0.083 μM for hCA XII). It is a valuable platform compound which can be prepared from malic acid. Coumalic acid can be used in the flavorings, fragrances and cosmetics industries, as polymer components, and pharmaceutical scaffolds. Coumalic acid displays antibronchial and antimalarial activity.
  • Inhibitor of human carbonic anhydrase (hCA) (KI = 0.073 μM for hCA IX; KI = 0.083 μM for hCA XII).
  • Valuable platform compound that can be prepared from malic acid.
  • Can be used in the flavorings, fragrances, and cosmetics industries.
  • Can be used as polymer components and pharmaceutical scaffolds.
  • Displays antibronchial and antimalarial activity.
Catalog No. 50-004-01966