Learn More
Medchemexpress LLC Coumaran (2,3-Dihydrobenzofuran) | 496-16-2 | 99.5% | C8H8O | 100 G
SDP
Supplier:  Medchemexpress LLC HY75247100G
Encompass_Preferred
Coumaran (2,3-Dihydrobenzofuran) is an AChE inhibitor that exhibits antileishmanial activity. It may develop antiparasitic abilities by activating macrophages and demonstrating immunomodulatory effects. Coumaran can also serve as a biopesticide.
  • AChE inhibitor
  • Antileishmanial activity
  • May acquire antiparasitic capabilities through activation of macrophages
  • Exerts immunomodulatory activity
  • Can be used as a biopesticide
  • Active against promastigote (IC50=1.042 μM) and amastigote (IC50=1.43 μM) forms
  • Not significantly cytotoxic to macrophages or erythrocytes at concentrations of 0.5-13 μM over 48 hours
  • Increases phagocyte and lysosomal activity and nitrite (NO2-) levels at concentrations of 0.5-13 μM over 24 hours
Catalog No. 50-004-34628