Learn More
Medchemexpress LLC Coumaran (2,3-Dihydrobenzofuran) | 496-16-2 | MFCD00005855 | 99.5% | C8H8O | 50 G
SDP
Supplier:  Medchemexpress LLC HY7524750G
Encompass_Preferred
Coumaran (2,3-Dihydrobenzofuran) is an AChE (Acetylcholinesterase) inhibitor that exhibits antileishmanial activity. It may acquire antiparasitic capabilities by activating macrophages and can also exert immunomodulatory activity. Coumaran is also identified as a biopesticide. For research use only, this product is not sold to patients.
  • AChE inhibitor
  • Antileishmanial activity against Leishmania amazonensis (IC50 promastigote=1.042 μM, amastigote=1.43 μM)
  • Immunomodulatory activity through macrophage activation
  • Functions as a biopesticide
  • Low cytotoxicity (0.5-13 μM for 48 hours)
  • Enhances phagocyte and lysosomal activity, and nitrite (NO2-) levels (0.5-13 μM for 24 hours)
Catalog No. 50-004-34377