Learn More
Medchemexpress LLC Coumberone | 878019-47-7 | 100.0% | 345.39 | C22H19NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD134110MG
Encompass_Preferred
Coumberone is a metabolic fluorogenic probe and isoform-selective substrate for aldo-keto reductase 1C (AKR1C) enzymes. It is enzymatically reduced to the fluorescent alcohol coumberol and is used to monitor AKR1C activity, induction, and substrate specificity in cellular assays.
  • Metabolic fluorogenic probe for AKR1C isoforms.
  • Enables selective optical readout and real-time imaging in cell-based assays.
  • High chemical purity suitable for research applications (99.95%).
  • Soluble in DMSO at ~16.67 mg/mL with recommended warming and ultrasonication.
  • Stable when stored at recommended temperatures for long-term use.
Catalog No. 50-002-97787