Learn More
Hach Company Counter-Hand-Tally
Supplier: Hach Company 1469600
For quick and accurate counting of colonies of bacteria and drops from pipets Tallies up to 9999 and resets easily to zero Mounted in 51 mm (2in ) case Aristo