Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD2004
Encompass_Preferred
CPC Ferruleless Polytube Inserts with Shut-Off Valve White Acetal Height 0 58 in 14 7 mm Overall Length 1 3 to 1 8 in 33 to 45 mm For Use With 1 6mm to 6 4mm tube Includes Shut-off valves PTF Valved In-Line Coupling Insert Maximum Pressure 120 psi
Catalog No. 50-313-1342