Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PLC22004
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow For Use With 6 4mm Buna-N Seal Max Operating Temperature 82 deg C 180 deg F Min Operating Temperature -40 deg C -40 deg F White Diameter Inner 0 25 mm
Catalog No. 50-313-1326