Learn More
COLDER PRODUCTS COMPANY COUPLER INSERT 25/PK
Supplier:  COLDER PRODUCTS COMPANY PMCD2204
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline Hose-Barb Inserts With Straight-Through Flow Product Line PMC Series Buna-N Seal For Use With 6 4mm For use in laboratory equipment medical and surgical instrumentation water transfer air systems and other applications
Catalog No. 50-313-1383