Learn More
COLDER PRODUCTS COMPANY COUPLING BODY LOCK 1/2 HB 250
Supplier:  COLDER PRODUCTS COMPANY MPXK17839
Encompass_Preferred
CPC Coupling Body with Lock Connector Amber tint Depth 1 96 in 49 8 mm Description USP Class VI ADCF Height 1 28 in 32 5 mm Polysulfone Pressure Max 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F
Catalog No. 50-313-1363
Only null left
Add to Cart
Request a Quote