Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPX17839 Quick-Disconnect Body, PS, 1/2"" ID; 5/Pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3131123
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-916