Learn More
COLDER PRODUCTS COMPANY CPC HFC35 Polysulfone Hose Barb Coupling Bodies In-line coupling Free floating EPDM seal options Vacuum to 125 psi 8 6 bar Valve Spring 316 Stainless Steel
Supplier:  COLDER PRODUCTS COMPANY HFCD17635
Encompass_Preferred
CPC HFC35 Polysulfone Hose Barb Coupling Bodies In-line coupling Free floating EPDM seal options Vacuum to 125 psi 8 6 bar Valve Spring 316 Stainless Steel
Catalog No. 50-313-1400