Learn More
Wilmad Labglass COUPLING FLEXIBLE 10MM 3/8IN
Supplier:  Wilmad Labglass LG9598120
Encompass
Coupling, Flexible, w/ PTFE Sleeve. Stirring Shaft Size: 19mm; Motor Shaft Size: 3/8 in. 4 in. length allows For compensation of misalignment. Rubber sleeve body provides quiet, vibration free running.
Catalog No. 50-800-333