Learn More
Ace Glass, Inc. COUPLING PTFE 25MM COUPLING PTFE 25MM
SDP
Supplier:  Ace Glass, Inc. 584049
Encompass_Preferred
25mm PTFE coupling for coupling threaded columns together, or coupling threaded columns to end fittings with a single o-ring seal for leak-tight engagement, with hand pressure and no significant size reduction in I.D. Supplied with two FETFE o-rings.
Catalog No. 50-978-500