Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD2302 Quick-Disconnect Hose Barb Elbow Insert; Valved, Delrin, 1/8"" Tube ID
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0636139
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-258