Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #0, 22x22 mm, 208 per oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7219811
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 208 per oz
Thickness: .08 - .13mm
Catalog No. NC1339805