Learn More
Electron Microscopy Sciences Coverglass Staining Rack, Stainless Steel
SDP
Supplier:  Electron Microscopy Sciences 72240
Encompass_Preferred
Coverglass staining rack made from polished stainless steel. Specially designed to be used with coplin jars. Holds five of 22 x 24 mm coverglass. Comes with a handle 5" (127 mm) long.
Catalog No. 50-949-581