Learn More
Med Vet International Ultrasound Probe Cover Eclipse 1-3/4 X 9-1/2 inch Polyisoprene Nonsterile For Use With Ultrasound Endocavity Probe, 38-01, 100/Box, 6 Boxes
Supplier:  Med Vet International 316682CS
Encompass_Preferred
Pre-gelled inside with Aquasonic 100 ultrasound transmission gel Only one probe cover is required for each procedure Fits most endocavity ultrasound probes
Catalog No. 50-283-1384