Learn More
Wilmad Labglass 4 Neck O-Ring Flange Reaction Vessel Cover 29/42 29/42
Supplier:  Wilmad Labglass LG8077104
Encompass_Preferred
Designed to be used with 2000mL, 3000mL, and 4000mL vessels (LG-8075, LG-8079A, LG-8079B and LG-8079C) . Flange OD is 6 5/8".
Catalog No. 50-805-692