Learn More
BIOSCIENCES TOOLS High Precision Glass Cover Slips, 18x18mm, 200/pk
Supplier:  BIOSCIENCES TOOLS CSHPNO1.518X18
Encompass
Cover glasses for high performance microscopes, made of chemically resistant borosilicate glass of the first hydrolytic class with precision thickness No. 1.5 (0.170 mm ± 0.005 mm), suitable for in-vitro diagnostic applications. Absolutely colorless, perfectly clear, suitable for fluorescence microscopy.
Catalog No. NC1832380