Learn More
HARVARD BIOSCIENCE INC COVER SLIP ROUND 1.5
Supplier:  HARVARD BIOSCIENCE INC 640715
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 11.High quality coverslip glass is essential for microscopy and imaging. Small diameter coverglass with diameters ranging from 3 to 8 mm are now available in convenient packages of 100. Round, square, and rectangular geometries. Tolerance 0.16-0.18 mm (160-180 m)
Catalog No. NC9560650