Learn More
Essendant COVERALLCHEMLG12/CSYL
Supplier:  Essendant ANS09813
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees If you or your employees are working in an environment where liquid and dry chemicals are present you need the right protection equipment You can trust the KleenGuard A70 Chemical Spray Protection Coveralls for comfort and protection They are made of a 1 5 mil polyethylene-coated fabric which is resistant to many water-based liquids greases and oils and liquid chemical splashes They feature a wick-away lining for enhanced comfort a liquid-resistant extra-long zipper and a taped sealable zipper flap The seamless front gives the wearer more protection in the primary exposure area and the REFLEX Coverall Design offers excellent fit and freedom of movement This model comes with a hood zipper front (with storm flap) and elastic wrists and ankles Making the right Personal Protection choice is difficult Perform with distinction wi
Catalog No. 50-000-22809