Learn More
Essendant COVERALLKLNGDA353XL
Supplier:  Essendant ANS38921
Encompass_Preferred
Ensure safe working conditions Lightweight and comfortable microporous film laminate provides an excellent liquid and particulate barrier Zipper front keeps the coveralls easy to put on and take off Reliable apparel for applications such as liquid handling pressure washing and general industrial tasks Zipper Front Apparel Type- Coverall Material(s)- Film Laminate Color(s)- White Color Family- White
Catalog No. 50-000-20405