Learn More
Essendant COVERALLLARGE25/CSWH
Supplier:  Essendant ANS38927
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits providing for safety and productivity for you and your employees When you re looking for an economical way to help protect your workers KleenGuard A35 Coveralls for non-hazardous particulate and light liquid splash protection are an excellent choice These disposable lightweight coveralls are made of microporous film-laminated material that helps protect against dry particulates and light splash KleenGuard A35 Coveralls feature a zippered front These safety coveralls provide protection during tasks like general industrial manufacturing and maintenance laboratory work and painting and help protect workers from dust dirt and light splashes KleenGuard A35 Coveralls meet design minimums in ANSI/ISEA 101-2014 (R2019) sizing standards In addition this product is eligible for recycling via RightCycle by Kimberly-Clark Professional (when used in non-hazardous applications as classified by federal state
Catalog No. 50-000-22813