Learn More
Essendant COVERKLNGDSHOEUNVSWH
Supplier:  Essendant ANS36885
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees KleenGuard A20 Shoe Covers are designed to move with you and your employees while helping protect against hazardous and non-hazardous dust These protective garments offer superior particle holdout coupled with unmatched comfort attributes These shoe covers are packaged for cleanrooms (double bagged) and feature seamless elastic sole toe and heel Shoe Covers Apparel Type- Shoe Cover Material(s)- MICROFORCE Barrier SMS Fabric Color(s)- White Color Family- White
Catalog No. 50-000-20404