Learn More
Electron Microscopy Sciences German Glass Coverslips #1, 5 mm
SDP
Supplier:  Electron Microscopy Sciences 7229001
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 5 mm
Catalog No. 50-192-9515