Learn More
Electron Microscopy Sciences Poly-D-Lysine German Glass Coverslips #1.5, 18 mm
SDP
Supplier:  Electron Microscopy Sciences 7229411
Encompass_Preferred
Sterilized by radiation, ready to use. Tested for cell cultures. User manual included.
Diameter: 18 mm
Catalog No. 50-192-9578