Learn More
Enterprise Technology Solutions Covidien Locking Wall Bracket, 5 Qt., Coated Metal
SDP
Supplier:  Enterprise Technology Solutions CVDSLWB10018X
Encompass_Preferred
Locking Wall Bracket is designed for use with 5 Quart In-Room Red Sharps Container and Clear Sharps Container. Lockable wall bracket provides added security. Coated metal wall bracket frees up valuable counter space and secures containers.
Catalog No. 50-008-31554