Learn More
TARGETMOL CHEMICALS INC COX-2-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T100325MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. COX-2-IN-2 is a selective and inducible COX2 inhibitor with an IC50 of 0.24 uM. COX-2-IN-1 is an anti-inflammatory compound with anti-inflammatory and analgesic activitiespurity: 98%
Catalog No. 50-258-1772