Learn More
TARGETMOL CHEMICALS INC COX-2-IN-6 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T620615MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. COX-2-IN-6 is a potent selective and orally available cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 0.84 uM and a Ki of 69 nM.COX-2-IN-6 inhibits COX-2-driven PGE2 synthesis with an IC50 of 0.60 uM.COX-2-IN-6 is used to prevent colorectal cancer. COX-2-IN-6 can be used to prevent colorectal cancer. purity: 99%
Catalog No. 50-257-9487