Learn More
Cayman Chemical COX Activity Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 76015196WELLS
Encompass_Preferred
The COX Activity Assay utilizes the peroxidase component of cyclooxygenases. The peroxidase activity is assayed colorimetrically by monitoring the appearance of oxidized TMPD at 590 nm.The assay can be used to measure COX activity (COX-1 and COX-2) in cell lysates, tissue homogenates, and purified enzyme preparations. The assay includes COX-1 and COX-2 specific inhibitors in order to distinguish between the two isozymes.
Catalog No. NC0399665