Learn More
Medchemexpress LLC CP-100356 hydrochloride | 142715-48-8 | MFCD00911858 | 99.1% | 597.10 g·mol⁻1 | C31H37ClN4O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY108347100MG
Encompass_Preferred
CP-100356 hydrochloride is a research-grade P-glycoprotein (MDR1) and BCRP inhibitor provided as the monohydrochloride salt for in vitro and preclinical studies. It is used to inhibit efflux transporters in cell-based assays to improve intracellular exposure and to evaluate transporter-mediated clearance. The compound is supplied as a solid powder and is available in small pack sizes suitable for screening and experimental use.
  • Research-grade P-gp and BCRP inhibitor for cellular assays.
  • Provided as the monohydrochloride salt in solid powder form.
  • High reported purity suitable for biochemical and cell-based studies.
  • Molecular weight 597.10 g·mol⁻1 and formula C31H37ClN4O6.
  • Available in small pack sizes and solution formats for flexible use.
Catalog No. 50-002-43661