Learn More
Medchemexpress LLC HY-19193 5mg Medchemexpress, CP-105696 CAS:158081-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY191935MG
Encompass_Preferred
Medchemexpress, HY-19193 5mg CP-105696 CAS:158081-99-3 CP-105696 is a potent and selective Leukotriene B4 Receptor antagonist, with an IC50 of 8.42 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2109