Learn More
Medchemexpress LLC Cp-113818 | 135025-12-6 | 99.7% | 470.8 g/mol | C24H42N2OS3 | 1 ML

Supplier: Medchemexpress LLC HY10544510MM 1ML
CP-113818 is a potent cholesterol acyltransferase (ACAT) inhibitor used in cell-based and animal studies to reduce Aβ production and amyloid pathology in Alzheimer's disease research. It is provided as a solid and as a ready-to-use 10 mM solution in DMSO for in vitro applications, with documented solubility and storage conditions for research use only.
- Potent ACAT inhibitory activity suitable for Alzheimer's disease research.
- High purity (99.7%).
- Molecular weight 470.8 g/mol.
- Soluble in DMSO at 100 mg/mL for in vitro use.
- Available as solution and multiple solid pack sizes.
- Storage conditions documented for powder and solutions.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.