Learn More
TARGETMOL CHEMICALS INC CP-312 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T270645MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CP-312 (Cardioprotectant 312) is an antioxidant defense response activator that protects hiPSC-CM viability by acting through induction of heme oxygenase-1 and targeting the antioxidant response network by inducing HMOX1 expression.CP-312 protects human iPSC-derived cardiomyocytes from oxidative stress. purity: 99%
Catalog No. 50-258-0652