Learn More
Abcam CP-31398 dihydrochloride, p53 stabilizing agent, 50MG
Supplier:  Abcam AB14164350MG
Encompass_Preferred
MW 435.4 Da, Purity >98%. p53 stabilizing agent. Promotes active conformation of p53 and restores wild-type conformation of mutant p53 in vitro. Able to induce cell cycle arrest and apoptosis. Chemopreventative activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6483