Learn More
Medchemexpress LLC CP-316819 | 186392-43-8 | 99.0% | 415.87 | 100 MG

Supplier: Medchemexpress LLC HY10861510MM/1ML
CP-316819 is a potent glycogen phosphorylase (GPase) inhibitor with antihyperglycemic effect. It has IC50 values of 17 nM against human skeletal muscle glycogen phosphorylase (huSMGPa) and 34 nM against liver glycogen phosphorylase (huLGPa). CP-316819 causes glycogen accumulation under normoglycemic conditions but permits glycogen utilization when glucose concentrations are low. It also prevents neuronal cell death and maintains brain electrical currents.
- Molecular weight: 415.87
- Formula: C21H22ClN3O4
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.