Learn More
Apexbio Technology LLC CP 339818 hydrochloride 50mg
SDP
Supplier:  Apexbio Technology LLC B672150
Encompass_Preferred
CP 339818 hydrochloride (CAS 478341-55-8) is a small-molecule antagonist targeting the Kv1 3 potassium channel It is designed to selectively inhibit Kv1 3-mediated ion conductance thereby modulating T-cell activation CP 339818 hydrochloride exerts its biological activity primarily through stabilization of the Kv1 3 channel s C-type inactivation state In in vitro studies CP 339818 hydrochloride demonstrates potent inhibition of Kv1 3 channels with an IC50 value of approximately 200 nM Based on these pharmacological properties CP 339818 hydrochloride holds research potential in the investigation of immune modulation mechanisms relevant to autoimmune and inflammatory disorders
Catalog No. 50-000-89771