Learn More
Medchemexpress LLC Cp-640186 hydrochloride | 591778-70-0 | MFCD28167776 | 99.7% | 522.08 g/mol | C30H36ClN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15259A10MG
Encompass_Preferred
CP-640186 hydrochloride is the hydrochloride salt of a research-grade acetyl-CoA carboxylase (ACC) inhibitor used in metabolic studies. It is orally active and cell-permeable, with reported low-nanomolar potency against ACC isoforms, and can stimulate muscle fatty acid oxidation. Intended for laboratory research use only.
  • Inhibits acetyl-CoA carboxylase with reported IC50s of 53 nM (ACC1) and 61 nM (ACC2).
  • Orally active and cell-permeable research compound.
  • High purity suitable for biochemical assays and cellular studies.
  • Solid, light yellow to pink appearance.
  • Molecular weight 522.08 g/mol; formula C30H36ClN3O3.
  • Available in multiple small vial sizes for dose-ranging experiments.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-86338