Learn More
Medchemexpress LLC HY-15259A 5mg Medchemexpress, CP-640186 (hydrochloride) CAS:591778-70-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15259A5MG
Encompass_Preferred
Medchemexpress, HY-15259A 5mg CP-640186 (hydrochloride) CAS:591778-70-0 CP-640186 hydrochloride is a potent and cell-permeable Acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 53 nM and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2 respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0599