Learn More
Selleck Chemical LLC CP-673451 10mM/1mL 343787-29-1
SDP
Supplier:  Selleck Chemical LLC S153610MM/1ML
Encompass_Preferred
CP-673451 is a selective inhibitor of PDGFR?/? with IC50 of 10 nM/1 nM in cell-free assays, exhibits >450-fold selectivity over other angiogenic receptors, has antiangiogenic and antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1361