Learn More
Apexbio Technology LLC CP-724714 537705-08-1 25mg
SDP
Supplier:  Apexbio Technology LLC A241225
Encompass_Preferred
CP-724714 (CAS 537705-08-1) is a small molecule inhibitor selectively targeting erbB2 (Her2) and EGFR kinases with notable potency against erbB2 kinase (IC50 10 3 nM) and considerably lower activity toward EGFR kinase (IC50 6 400 2 100 nM) In cellular assays CP-724714 induces G1 cell-cycle arrest and decreases erbB2 phosphorylation in Her2-overexpressing BT-474 breast cancer cells In animal xenograft models (BT-474 MDA-MB-453) it reduces erbB2 receptor phosphorylation and downstream signaling resulting in tumor growth inhibition and apoptosis induction CP-724714 advanced into phase I clinical trials for Her2-driven breast cancer research
Catalog No. 50-000-62998