Learn More
Medchemexpress LLC CP-724714 | 383432-38-0 | 98.7% | 469.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14674
Encompass_Preferred
CP-724714 is a potent, selective, and orally active ErbB2 (HER2) tyrosine kinase inhibitor. It effectively inhibits ErbB2 receptor autophosphorylation in intact cells and demonstrates antitumor activities. With an IC50 of 10 nM for ErbB2, it shows significant selectivity against EGFR kinase.
  • Potent and selective ErbB2 (HER2) tyrosine kinase inhibitor
  • Orally active
  • Inhibits ErbB2 receptor autophosphorylation
  • Exhibits antitumor activities
  • Induces G1 cell cycle block in ErbB2-overexpressing cells
  • Demonstrates favorable nonclinical toxicity
Catalog No. 50-003-63334