Learn More
Apexbio Technology LLC CP 94253 hydrochloride 845861-39-4 10mg
SDP
Supplier:  Apexbio Technology LLC B668510
Encompass_Preferred
CP 94253 hydrochloride (CAS 845861-39-4) is a highly selective agonist of the 5-hydroxytryptamine 1B (5-HT1B) receptor In radioligand binding assays it demonstrates potent affinity for the 5-HT1B receptor with a Ki value of 2 nM By specifically activating 5-HT1B receptors CP 94253 hydrochloride modulates serotonergic neurotransmission and has been shown to exhibit activity within the central nervous system in in vivo studies This compound is widely utilized in neuroscience research to investigate the function of 5-HT1B receptors and their involvement in neuropsychiatric and neurological disorders
Catalog No. 50-000-91026