Learn More
Cayman Chemical CP 945 598hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1195510 MG
Encompass_Preferred
A selective CB1 receptor antagonist (Ki = 0.7 nM); reverses cannabinoid agonist-mediated responses
Catalog No. 50-295-4055