Learn More
Cayman Chemical CP 945 598hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 119555 MG
Encompass_Preferred
A selective CB1 receptor antagonist (Ki = 0.7 nM); reverses cannabinoid agonist-mediated responses
Catalog No. 50-295-3304