Learn More
Andwin Scientific CP CRYOMARKERS BLACK 10/PK
SDP
Supplier:  Andwin Scientific 0996400
Encompass_Preferred
The Cole-Parmer lab marker feature fine point. This product can write on cold, wet surfaces. Its ink dries quickly and stays permanently on plastic. When used on glass, metal, or porcelain, the ink can be washed off but will not bleed through upon encounter with water or alcohol. This is sold in a pack of 10 markers.
Catalog No. NC9622469