Learn More
Enzo Life Sciences Cyclophosphamide . monohydrate (1g). CAS: 6055-19-2
SDP
Supplier:  Enzo Life Sciences ALX400051G001
Encompass_Preferred
Alkylates DNA, forming DNA-DNA cross-links that result in inhibition of DNA synthesis and cell death. Interestingly the repair enzyme O6-alkylguanine-DNA alkyltransferase, long thought to be involved with resistance to methylating and chloroethylating agents, may also contribute to resistance to the cytotoxic and mutagenic effects of cyclophosphamide. Alternative name: 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan. Purity: ≥97% (HPLC). Solubility: Soluble in water; slightly soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9089